Learn More
rac-trans-4-Defluoro-4-methoxy Paroxetine Hydrochloride, TRC
Specifications
Specifications
| CAS | 127017-74-7 |
| Molecular Formula | C20 H23 N O4 . Cl H |
| Quantity | 10 mg |
| Synonym | Piperidine, 3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-methoxyphenyl)-, hydrochloride (1:1), (3R,4S)-rel-, Piperidine, 3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-methoxyphenyl)-, hydrochloride, trans- (9CI), Piperidine, 3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-methoxyphenyl)-, hydrochloride, trans-(±)-, SB-242670-A, Ph Eur Paroxetine Hydrochloride Impurity B, p-Methoxy Paroxetine HCl, USP Paroxetine Related Compound A |
| InChI Formula | InChI=1S/C20H23NO4.ClH/c1-22-16-4-2-14(3-5-16)18-8-9-21-11-15(18)12-23-17-6-7-19-20(10-17)25-13-24-19;/h2-7,10,15,18,21H,8-9,11-13H2,1H3;1H/t15-,18-;/m0./s1 |
| SMILES | Cl.COc1ccc(cc1)[C@@H]2CCNC[C@H]2COc3ccc4OCOc4c3 |
| IUPAC Name | (3S,4R)-3-(1,3-benzodioxol-5-yloxymethyl)-4-(4-methoxyphenyl)piperidine;hydrochloride |
| Molecular Weight (g/mol) | 377.86 |
| Formula Weight | 377.1394 |
| Percent Purity | >95% |
| Show More |