Learn More
rac-Propanolol, TRC
Product Code. 18060320
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
100mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18060320 50 mg 50mg
18051410 5 mg 5mg
18032820 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18060320 Supplier Toronto Research Chemicals Supplier No. TRCP84001350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 525-66-6
Molecular Formula C16 H21 N O2
Molecular Weight (g/mol) 259.34
Synonym 1-[(1-Methylethyl)amino]-3-(1-naphthalenyloxy)-2-propanol, (±)-Propranolol, 1-(1-Naphthyloxy)-3-(isopropylamino)-2-propanol, 1-(Isopropylamino)-3-(1-naphthyloxy)-2-propanol, 3-(Naphth-1-yloxy)-1-(isopropylamino)propan-2-ol, AY 64043, Betalong, DL-Propranolol, Euprovasin, Innopran XL, Propranolol, Proprasylyt, Racemic propranolol, Reducor, dl-Propranolol, β-Propranolol, rac-Propanolol, 1[(1-Methylethyl)amino]-3-(1-naphthalenyloxy)-2-propanol
IUPAC Name 1-(isopropylamino)-3-(1-naphthyloxy)propan-2-ol
SMILES CC(C)NCC(O)COc1cccc2ccccc12
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C16H21NO2/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16/h3-9,12,14,17-18H,10-11H2,1-2H3
Formula Weight 259.1572
Percent Purity >95%
Chemical Name or Material Propranolol
Grade HPLC