missing translation for 'onlineSavingsMsg'
Learn More

rac-N-Benzyl Formoterol (Mixture of Diastereomers), TRC

Product Code. 30136597
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136597 100 mg 100mg
30007995 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136597 Supplier Toronto Research Chemicals Supplier No. TRCB279790100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

InChI Formula InChI=1S/C26H30N2O4/c1-19(14-20-8-11-23(32-2)12-9-20)28(16-21-6-4-3-5-7-21)17-26(31)22-10-13-25(30)24(15-22)27-18-29/h3-13,15,18-19,26,30-31H,14,16-17H2,1-2H3,(H,27,29)
IUPAC Name N-[5-[2-[benzyl-[1-(4-methoxyphenyl)propan-2-yl]amino]-1-hydroxyethyl]-2-hydroxyphenyl]formamide
Molecular Formula C26H30N2O4
Molecular Weight (g/mol) 434.53
SMILES OC(CN(CC1=CC=CC=C1)C(CC2=CC=C(OC)C=C2)C)C3=CC=C(O)C(NC=O)=C3
Synonym rac-N-[2-Hydroxy-5-[1-hydroxy-2-[[2-(4-methoxyphenyl)-1-methylethyl](phenylmethyl)amino]ethyl]phenyl]formamide
Chemical Name or Material rac-N-Benzyl Formoterol (Mixture of Diastereomers)

Specifications

Analyte or Component Names rac-N-Benzyl Formoterol (Mixture of Diastereomers)
Shipping Condition Room Temperature
Solution Type Neat
Industry Type Pharmaceutical
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.