missing translation for 'onlineSavingsMsg'
Learn More

rac-N-Benzyl Formoterol (Mixture of Diastereomers), TRC

Product Code. 30007995
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007995 10 mg 10mg
30136597 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007995 Supplier Toronto Research Chemicals Supplier No. TRCB27979010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

InChI Formula InChI=1S/C26H30N2O4/c1-19(14-20-8-11-23(32-2)12-9-20)28(16-21-6-4-3-5-7-21)17-26(31)22-10-13-25(30)24(15-22)27-18-29/h3-13,15,18-19,26,30-31H,14,16-17H2,1-2H3,(H,27,29)
IUPAC Name N-[5-[2-[benzyl-[1-(4-methoxyphenyl)propan-2-yl]amino]-1-hydroxyethyl]-2-hydroxyphenyl]formamide
Molecular Formula C26H30N2O4
Molecular Weight (g/mol) 434.53
SMILES OC(CN(CC1=CC=CC=C1)C(CC2=CC=C(OC)C=C2)C)C3=CC=C(O)C(NC=O)=C3
Synonym rac-N-[2-Hydroxy-5-[1-hydroxy-2-[[2-(4-methoxyphenyl)-1-methylethyl](phenylmethyl)amino]ethyl]phenyl]formamide
Chemical Name or Material rac-N-Benzyl Formoterol (Mixture of Diastereomers)

Especificaciones

Analyte or Component Names rac-N-Benzyl Formoterol (Mixture of Diastereomers)
Shipping Condition Room Temperature
Solution Type Neat
Industry Type Forensics
Quantity 10 mg
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.