Learn More
(Rac)-IBT6A, MedChemExpress
Product Code. 16471677
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471677 50 mg 50mg
16458217 5 mg 5mg
16416086 100 mg 100mg
16415508 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16471677 Supplier Medchem Express Supplier No. HY13036
MedChemExpress (Rac)-IBT6A is a racemate of IBT6A. IBT6A is an impurity of Ibrutinib. IBT6A can be used in synthesis of IBT6A Ibrutinib dimer and IBT6A adduct. Ibrutinib is a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM.
(Rac)-IBT6A is a racemate of IBT6A. IBT6A is an impurity of Ibrutinib. IBT6A can be used in synthesis of IBT6A Ibrutinib dimer and IBT6A adduct. Ibrutinib is a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM.
Specifications
Chemical Name or Material (Rac)-IBT6A
CAS 1412418-47-3
Color White
Purity Grade Notes Research
Molecular Formula C22H22N6O
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (129.38 mM; Need ultrasonic)
SMILES NC1=C2C(N(C3CNCCC3)N=C2C4=CC=C(OC5=CC=CC=C5)C=C4)=NC=N1
Molecular Weight (g/mol) 386.45
Formula Weight 386.45
Percent Purity 98.18%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation