missing translation for 'onlineSavingsMsg'
Learn More

rac Alminoprofen, TRC

Product Code. 30328495
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30328495 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328495 Supplier Toronto Research Chemicals Supplier No. TRCA57515025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (±)-Alminoprofen
CAS 39718-89-3
Molecular Formula C13 H17 N O2
Quantity 25 mg
Synonym Benzeneacetic acid, α-methyl-4-[(2-methyl-2-propen-1-yl)amino]-, Benzeneacetic acid, α-methyl-4-[(2-methyl-2-propenyl)amino]- (9CI), α-Methyl-4-[(2-methyl-2-propen-1-yl)amino]benzeneacetic acid, (±)-Alminoprofen, Alminoprofen, EB 382, Minalfene
InChI Formula InChI=1S/C13H17NO2/c1-9(2)8-14-12-6-4-11(5-7-12)10(3)13(15)16/h4-7,10,14H,1,8H2,2-3H3,(H,15,16)
SMILES CC(C(=O)O)c1ccc(NCC(=C)C)cc1
IUPAC Name 2-[4-(2-methylprop-2-enylamino)phenyl]propanoic acid
Molecular Weight (g/mol) 219.28
Formula Weight 219.1259
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.