missing translation for 'onlineSavingsMsg'
Learn More

Rabeprazole-d3 Sodium Salt, TRC

Product Code. 30411787
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30411787 1 mg
30410899 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30411787 Supplier Toronto Research Chemicals Supplier No. TRCR0705031MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C18H17D3N3NaO3S
Molecular Weight (g/mol) 384.44
Synonym 2-[[[(4-(3-Methoxy-d3)propoxy)-3-methyl-2-pyridinyl]methyl]sulfinyl]-1H-benzimidazole Sodium Salt
SMILES CC(C(OCCCOC([2H])([2H])[2H])=CC=N1)=C1CS(C(N2[Na])=NC3=C2C=CC=C3)=O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C18H20N3O3S.Na/c1-13-16(19-9-8-17(13)24-11-5-10-23-2)12-25(22)18-20-14-6-3-4-7-15(14)21-18;/h3-4,6-9H,5,10-12H2,1-2H3;/q-1;+1/i2D3;
Chemical Name or Material Rabeprazole - d3 Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.