missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYN1 Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native SYN1.
Brand: Abnova PAB9702.10ug
This item is not returnable.
View return policy
Specifications
| SYN1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against native SYN1. | |
| Lyophilized from 5mM ammonium bicarbonate | |
| MGC152080 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 281510 | |
| Store at -20°C on dry atmosphere. After reconstitution with PBS store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Frozen sections) (1:2000) Immunofluorescence (1:2000) Immunoprecipitation (1 ug/200 ug lysate ) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| SYN1 | |
| SYN1 | |
| Native purified bovine SYN1. | |
| 10 μg | |
| Primary | |
| Bovine, Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction