Learn More
EMCN Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of EMCN.
Brand: Abnova PAB17468.100ug
Description
EMCN is a mucin-like sialoglycoprotein that interferes with the assembly of focal adhesion complexes and inhibits interaction between cells and the extracellular matrix (Kinoshita et al., 2001 [PubMed 11418125]).[supplied by OMIM
Specifications
| EMCN | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of EMCN. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| Q9ULC0 | |
| EMCN | |
| A synthetic peptide corresponding to internal of human EMCN. | |
| RUO | |
| 51705 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunofluorescence | |
| Unconjugated | |
| Immunofluorescence (1:500-1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user. | |
| EMCN | |
| EMCN2/MUC14 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total EMCN protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only