Learn More
BSN, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant BSN.
Brand: Abnova PAB22825.100uL
Description
Neurotransmitters are released from a specific site in the axon terminal called the active zone, which is composed of synaptic vesicles and a meshwork of cytoskeleton underlying the plasma membrane. The protein encoded by this gene is thought to be a scaffolding protein involved in organizing the presynaptic cytoskeleton. The gene is expressed primarily in neurons in the brain. A similar gene product in rodents is concentrated in the active zone of axon terminals and tightly associated with cytoskeletal structures, and is essential for regulating neurotransmitter release from a subset of synapses. [provided by RefSeq
Sequence: AQEHTFLATATTVSITMASSVFMAQQKQPVVYGDPYQSRLDFGQGGGSPVCLAQVKQVEQAVQTAPYRSGPRGRPREAKFARYNLPNQVAPLASpecifications
| BSN | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant BSN. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9UPA5 | |
| BSN | |
| Recombinant protein corresponding to amino acids of human BSN. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| BSN | |
| ZNF231 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 8927 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only