missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB7 Recombinant Rabbit Monoclonal Antibody (5A10A4)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535163
This item is not returnable.
View return policy
Description
Immunogen sequence: IQASPRDPEN FPFVVLGNKI DLENRQVATK RAQAWCYSKN NIPYFETSAK EAINVEQAFQ TIARNALKQE TEVELYNEFP EPIKLDKNDR AKASAESCSC.
RAB family members are small, RAS-related GTP-binding proteins that are important regulators of vesicular transport. Each RAB protein targets multiple proteins that act in exocytic / endocytic pathways. This gene encodes a RAB family member that regulates vesicle traffic in the late endosomes and also from late endosomes to lysosomes. This encoded protein is also involved in the cellular vacuolation of the VacA cytotoxin of Helicobacter pylori. Mutations at highly conserved amino acid residues in this gene have caused some forms of Charcot-Marie-Tooth (CMT) type 2 neuropathies.
Specifications
| RAB7 | |
| Recombinant Monoclonal | |
| 0.6 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P09527, P51149, P51150 | |
| Rab7, RAB7A | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| 5A10A4 | |
| Unconjugated | |
| RAB7A | |
| GTP-binding protein (rab7); hypothetical protein LOC509970; PRO2706; RAB7; rab-7; RAB7, member RAS oncogene family; Rab7a; RAB7A, member RAS oncogene family; Ras-associated protein RAB7; ras-related protein BRL-RAS; Ras-related protein p23; ras-related protein Rab-7a | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 110315128, 19349, 29448, 7879 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido