missing translation for 'onlineSavingsMsg'
Learn More

RAB5B Polyclonal Antibody
GREENER_CHOICE

Product Code. 15939664
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15939664 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15939664 Supplier Invitrogen Supplier No. PA566986

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

RAB5B Polyclonal Antibody

RAB5B Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB5B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAB5B
Gene Accession No. P61020
Gene Alias C030027M18Rik; RAB5A member RAS oncogene family; RAB5B; RAB5B, member RAS oncogene family; ras-related protein Rab-5B; zgc:76978
Gene Symbols RAB5B
Host Species Rabbit
Immunogen Recombinant Human RAB5B. Recombinant protein control fragment (Product #RP-105447).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5869
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.