missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAB2B Polyclonal Antibody

Product Code. 15769961 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15769961 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15769961 Supplier Invitrogen™ Supplier No. PA562165

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GIKIGPQQSI STSVGPSASQ RNSRDIGSNS GCC Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 76%.

Members of the Rab protein family are nontransforming monomeric GTP-binding proteins of the Ras superfamily that contain 4 highly conserved regions involved in GTP binding and hydrolysis. Rab proteins are prenylated, membrane-bound proteins involved in vesicular fusion and trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB2B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAB2B
Gene Accession No. Q8WUD1
Gene Alias 1500012D09Rik; 4930528G15Rik; A230002G14; D530043M21Rik; GTP-binding protein RAB2B; Rab2b; RAB2B, member RAS oncogene family; RAS family, member RAB2B; ras-related protein Rab-2B
Gene Symbols RAB2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RAB2B. Recombinant protein control fragment (Product #RP-101282).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84932
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.