missing translation for 'onlineSavingsMsg'
Learn More

RAB11FIP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16824405 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16824405 20 μL 20µL
16814405 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16824405 Supplier Proteintech Supplier No. 258431AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Proteins of the large Rab GTPase family have regulatory roles in the formation, targeting, and fusion of intracellular transport vesicles. RAB11FIP3 is one of many proteins that interact with and regulate Rab GTPases (Hales et al., 2001 ).

Specifications

Antigen RAB11FIP3
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RAB11FIP3
Gene Accession No. O75154, Q8CHD8
Gene Alias ARFO1, Arfophilin 1, Eferin, FIP3 Rab11, KIAA0665, MU MB 17.148, Rab11 FIP3, RAB11FIP3
Gene Symbols RAB11FIP3
Host Species Rabbit
Immunogen RAB11FIP3 Fusion Protein Ag22937
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 215445, 303002, 9727
Target Species Mouse, Rat, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.