missing translation for 'onlineSavingsMsg'
Learn More

R406, MedChemExpress

Product Code. 16425746
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16425746

Brand: Medchem Express HY12067

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress R406 is an orally available and competitive Syk/FLT3 inhibitor for ATP binding with a Ki of 30 nM, potently inhibits Syk kinase activity in vitro with an IC50 of 41 nM, measured at an ATP concentration corresponding to its Km value. R406 reduces immune complex-mediated inflammation. R406 also inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM).

R406 is an orally available and competitive Syk/FLT3 inhibitor for ATP binding with a Ki of 30 nM, potently inhibits Syk kinase activity in vitro with an IC50 of 41 nM, measured at an ATP concentration corresponding to its Km value. R406 reduces immune complex-mediated inflammation. R406 also inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM).

Specifications

Chemical Name or Material R406
CAS 841290-81-1
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C28H29FN6O8S
Quantity 100 mg
Solubility Information DMSO : ≥ 61 mg/mL (97.04 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES COC1=CC(NC2=NC(NC3=NC(N4)=C(C=C3)OC(C)(C)C4=O)=C(C=N2)F)=CC(OC)=C1OC.OS(C5=CC=CC=C5)(=O)=O
Molecular Weight (g/mol) 628.63
Formula Weight 628.63
Percent Purity 96.67%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.