Learn More
R112, MedChemExpress
Product Code. 16499587
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16499587
Brand: Medchem Express HY16420
MedChemExpress R112 is an ATP-competitive inhibitor of Syk kinase with a Ki of 96 nM. R112 inhibits Syk kinase activity with an IC50 of 226 nM.
R112 is an ATP-competitive inhibitor of Syk kinase with a Ki of 96 nM. R112 inhibits Syk kinase activity with an IC50 of 226 nM.
Specifications
Chemical Name or Material R112
CAS 575474-82-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H13FN4O2
Quantity 5 mg
Solubility Information DMSO : 55 mg/mL (176.11 mM; Need ultrasonic)
SMILES FC1=CN=C(NC2=CC(O)=CC=C2)N=C1NC3=CC(O)=CC=C3
Molecular Weight (g/mol) 312.3
Formula Weight 312.3
Percent Purity 99.23%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation