Learn More
(R,S)-N-Acetyl-S-[1-(hydroxymethyl)-2-propen-1-yl)-L-cysteine + (R,S)-N-Acetyl-S-(2-hydroxy-3-buten-1-yl)-L-cysteine, TRC
Product Code. 30011888
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011888 500 mg 500mg
30012749 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30011888 Supplier Toronto Research Chemicals Supplier No. TRCA179005500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2762993-90-6
Molecular Formula C9H15NO4S
Molecular Weight (g/mol) 233.29
Synonym MHBMA
IUPAC Name N-acetyl-S-(1-hydroxybut-3-en-2-yl)-L-cysteine
SMILES C=CC(CSC[C@@H](C(O)=O)NC(C)=O)O.C=CC(SC[C@@H](C(O)=O)NC(C)=O)CO
Specifications
InChI Formula InChI=1S/C9H15NO4S/c1-3-7(12)4-15-5-8(9(13)14)10-6(2)11/h3,7-8,12H,1,4-5H2,2H3,(H,10,11)(H,13,14)/t7?,8-/m0/s1
Quantity 500 mg
Formula Weight 233.07
Shipping Condition Room Temperature
Chemical Name or Material N-Acetyl-S-(2-hydroxy-3-buten-1-yl)-L-cysteine + N-Acetyl-S-[1-(hydroxymethyl)-2-propen-1-yl]-L-cysteine