missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-BD-AcAc 2, MedChemExpress

Product Code. p-200101272
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16416746

Brand: Medchem Express HY15344

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (R,R)-BD-AcAc 2 ((R,R)-Ketone Ester), a ketone monoester, can be used as a source of oral nutritional ketones.

(R,R)-BD-AcAc 2 can elevate plasma levels of β-hydroxybutyrate and acetoacetate, blood glucose, blood Na+ levels and blood creatinine levels after oral administration in mice. (R,R)-BD-AcAc 2 can partly prevent muscle weakness in septic mice. (R,R)-BD-AcAc 2 has potential to improve exercise performance and endurance in animal body. (R,R)-BD-AcAc 2 can also be used to research Parkinson’s disease or diabetes.

Specifications

Chemical Name or Material (R,R)-BD-AcAc 2
Physical Form Viscous Liquid
CAS 1208313-97-6
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C8H16O4
For Use With (Application) Neuroscience-Neurodegeneration
Synonym (R,R)-Ketone Ester
Solubility Information H2O : 100 mg/mL (567.50 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (567.50 mM; Need ultrasonic)
SMILES C[C@@H](O)CC(OCC[C@H](O)C)=O
Molecular Weight (g/mol) 176.21
Formula Weight 176.21
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.