Learn More
(R)-Ketorolac Glucuronide, TRC
Specifications
Specifications
| Chemical Name or Material | (R)-Ketorolac Glucuronide |
| CAS | 233666-91-6 |
| Molecular Formula | C21H21NO9 |
| Quantity | 5 mg |
| Synonym | 1-[(1R)-5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylate]-β-D-glucopyranuronic Acid |
| InChI Formula | InChI=1S/C21H21NO9/c23-14(10-4-2-1-3-5-10)13-7-6-12-11(8-9-22(12)13)20(29)31-21-17(26)15(24)16(25)18(30-21)19(27)28/h1-7,11,15-18,21,24-26H,8-9H2,(H,27,28)/t11-,15+,16+,17-,18+,21+/m1/s1 |
| SMILES | O=C(C1=CC=CC=C1)C2=CC=C3N2CC[C@H]3C(O[C@@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O)C(O)=O)=O |
| IUPAC Name | (2S,3S,4S,5R,6S)-6-(((R)-5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carbonyl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic acid |
| Molecular Weight (g/mol) | 431.39 |
| Formula Weight | 431.12 |