missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-JQ1 Enantiomer, MedChemExpress

Product Code. 16498207
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498207 5 mg 5mg
16466076 100 mg 100mg
16455498 10 mg 10mg
16411677 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498207 Supplier Medchem Express Supplier No. HY13030A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (R)-(-)-JQ1 Enantiomer is the stereoisomer of (+)-JQ1. (+)-JQ1 potently decreases expression of both BRD4 target genes, whereas (R)-(-)-JQ1 Enantiomer has no effect.

(R)-(-)-JQ1 Enantiomer is the stereoisomer of (+)-JQ1. (+)-JQ1 potently decreases expression of both BRD4 target genes, whereas (R)-(-)-JQ1 Enantiomer has no effect.

Specifications

Chemical Name or Material (R)-(-)-JQ1 Enantiomer
Physical Form Solid
CAS 1268524-71-5
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C23H25ClN4O2S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 100 mg/mL (218.82 mM)
SMILES O=C(C[C@H]1N=C(C2=C(N3C1=NN=C3C)SC(C)=C2C)C4=CC=C(C=C4)Cl)OC(C)(C)C
Molecular Weight (g/mol) 456.99
Formula Weight 456.99
Percent Purity 98.63%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.