Learn More
R&D Systems™ Caspase Inhibitor Control Z-FA-FMK
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
Cell permeable fluoromethyl ketone (FMK)-derivatized peptides act as effective irreversible Caspase inhibitors with no cytotoxic effects and, therefore, are useful tools for studying Caspase activity.
- Treated cells can then be assayed for evidence of apoptosis inhibition by examining either whole cells or cell lysates using standard apoptosis assays
- Reconstitute in highly pure (ACS grade) DMSO fore use
Specifications
Specifications
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 5 mg |
| Product Type | Caspase Inhibitor Control Z-FA-FMK, 386Da |