Learn More
Tocris Bioscience™ R 7050
Inhibitor of TNF-α receptor 1 signaling
Brand: Tocris Bioscience™ 5432/50
This item is not returnable.
View return policy
Description
R 7050 is an inhibitor of TNF-α receptor 1 (TNFR1) signaling; blocks TNFR1 association with TRADD and RIP1. Inhibits TNF-α-induced NF-κB and MAPK signaling pathway activation. Attenuates intracerebral hemorrhage-induced neurovascular injury in mice.Specifications
| >98% | |
| 50 mg |
Product Title
Spot an opportunity for improvement?
For Research Use Only