Learn More
Tocris Bioscience™ R 7050
GREENER_CHOICE
Inhibitor of TNF-α receptor 1 signaling
Brand:  Tocris Bioscience™ 5432/50
Encompass
Product Code. 15786826
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
R 7050 is an inhibitor of TNF-α receptor 1 (TNFR1) signaling; blocks TNFR1 association with TRADD and RIP1. Inhibits TNF-α-induced NF-κB and MAPK signaling pathway activation. Attenuates intracerebral hemorrhage-induced neurovascular injury in mice.
Specifications
Specifications
>98%
50 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions