missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-Amino-N-benzyl-3-methoxypropionamide, TRC

Product Code. 30004362
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004362 100 mg 100mg
30134108 250 mg 250mg
30138660 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004362 Supplier Toronto Research Chemicals Supplier No. TRCA598000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 196601-69-1
Formula Weight 208.1212
InChI Formula InChI=1S/C11H16N2O2/c1-15-8-10(12)11(14)13-7-9-5-3-2-4-6-9/h2-6,10H,7-8,12H2,1H3,(H,13,14)/t10-/m1/s1
IUPAC Name (2R)-2-amino-N-benzyl-3-methoxypropanamide
Molecular Formula C11 H16 N2 O2
Molecular Weight (g/mol) 208.26
Percent Purity >95%
SMILES COC[C@@H](N)C(=O)NCc1ccccc1
Synonym Propanamide, 2-amino-3-methoxy-N-(phenylmethyl)-, (R)-, (2R)-2-Amino-3-methoxy-N-(phenylmethyl)propanamide, (2R)-2-Amino-3-methoxy-N-(phenylmethyl)propanamide, (2R)-2-Amino-N-benzyl-3-methoxypropanamide, (R)-2-Amino-N-benzyl-3-methoxypropionamide
Chemical Name or Material (R)-2-Amino-N-benzyl-3-methoxypropionamide
Grade HPLC
Show More Show Less

Specifications

Analyte or Component Names (R)-2-Amino-N-benzyl-3-methoxypropionamide
Shipping Condition Room Temperature
Solution Type Neat
Industry Type Forensics
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.