missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-Acetoxy-3-((trimethylsilyl)oxy)propyl Oleate, TRC

Product Code. 30328003
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30328003 100 mg 100mg
30319215 250 mg 250mg
30318321 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30328003 Supplier Toronto Research Chemicals Supplier No. TRCA187938100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-2-Acetoxy-3-((trimethylsilyl)oxy)propyl Oleate
Analyte or Component Names (R)-2-Acetoxy-3-((trimethylsilyl)oxy)propyl Oleate
Shipping Condition Dry ice
Molecular Formula C26 H50 O5 Si
InChI Formula InChI=1S/C26H50O5Si/c1-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-26(28)29-22-25(31-24(2)27)23-30-32(3,4)5/h13-14,25H,6-12,15-23H2,1-5H3/b14-13-/t25-/m1/s1
Solution Type Neat
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OC[C@H](CO[Si](C)(C)C)OC(=O)C
IUPAC Name [(2R)-2-acetyloxy-3-trimethylsilyloxypropyl] (Z)-octadec-9-enoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 470.758
Quantity 100 mg
Formula Weight 470.343
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.