missing translation for 'onlineSavingsMsg'
Learn More

R-10015, MedChemExpress

Product Code. 16471157
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471157 50 mg 50mg
16484908 10 mg 10mg
16407667 5 mg 5mg
16405696 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16471157 Supplier Medchem Express Supplier No. HY120097

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress R-10015, a broad-spectrum antiviral compound for HIV infection, acts as a potent and selective inhibitor of LIM domain kinase (LIMK) and binds to the ATP-binding pocket, with an IC50 of 38 nM for human LIMK1.

R-10015, a broad-spectrum antiviral compound for HIV infection, acts as a potent and selective inhibitor of LIM domain kinase (LIMK) and binds to the ATP-binding pocket, with an IC50 of 38 nM for human LIMK1.

Specifications

Chemical Name or Material R-10015
Physical Form Solid
CAS 2097938-51-5
Color Pink
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H19ClN6O2
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : 62.5 mg/mL (152.12 mM; Need ultrasonic)
SMILES O=C(C1=CC=C2NC(C3CCN(C4=C5C(NC=C5Cl)=NC=N4)CC3)=NC2=C1)OC
Molecular Weight (g/mol) 410.86
Formula Weight 410.86
Percent Purity 98.89%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.