missing translation for 'onlineSavingsMsg'
Learn More

Quinine, 95%, MP Biomedicals™

Product Code. 11456172 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
Unit Size:
100g
10g
25g
50g
This item is not returnable. View return policy

Product Code. 11456172

Brand: MP Biomedicals 0210279250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 130-95-0
Molecular Formula C20H24N2O2
Molecular Weight (g/mol) 324.42
MDL Number MFCD00198096
InChI Key LOUPRKONTZGTKE-ZCXAXGJJNA-N
Synonym quinine, chinin, chinine, --quinine, qualaquin, 6'-methoxycinchonidine, 8s,9r-quinine, chininum, quinine sulfate, quinina
PubChem CID 129316724
IUPAC Name (R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol
SMILES [H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C

Specifications

Quantity 50 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.