missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ QuicKit Control for Human/Cynomolgus Monkey PD-L1/B7-H1
Specifications
Specifications
| Quantity | 1 Ea. |
| Species | Human, Cynomolgus Monkey |
| Binding Target | PD-L1 |
| Gene ID (Entrez) | 29126 |
| Synonym | Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?