Learn More
Quickfit™ Swan Neck Adapter
Click to view available options
Cone Size:
19/26
24/29
Unit Size:
Each
Description
Has screwthread joint to accept thermometers and tubes
- Available in different socket, cone, and thread sizes
Specifications
Specifications
| Cone Size | 19/26 |
| For Use With (Application) | Ideal for securing thermometers, gas inlet tubes |
| Material | Borosilicate Glass |
| Product Type | Swan neck adapter |
| Thread Size | 13 |