Learn More
Quickfit™ Pressure Equalizing Funnel with Rotaflo™ Stopcock
Product Code. 12024453
Click to view available options
Capacity (Metric):
250 mL
Cone Size:
19/26
24/29
Joint Number:
24/29
Unit Size:
Each
Product Code. 12024453
Brand: Quickfit DE250/33
Only null left
Add to basket
Add to basket
For use when liquids have to be introduced into vessels under vacuum
  • Complies with ISO 4800
  • Can also be used with Soxhlets as solvent traps to facilitate recovery at the end of an extraction
  • With GP Rotaflo™ stopcocks having chemically resistant PTFE keys
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 250 mL
Material Borosilicate Glass
Certifications/Compliance ISO 4800
Cone Size 24/29
For Use With (Application) Used when liquids have to be introduced into vessels under vacuum, Soxhlets as solvent trap
Joint Number 24/29
Quantity 1PK
Stopcock GP Rotaflo™; 3mm
Product Type Pressure-equalizing Funnel