Learn More
Quickfit™ Female Spherical Ball Joint
Description
For use with male ball joints MS:
- Female cup joints precision manufactured to the highest standards
- Spherical joints are manufactured with an approximate shank length of 100mm
Specifications
Specifications
| Type | Female Spherical Ball Joint |
| Joint Size | S19 |
| Thickness (English) Wall | 0.06in. |
| Thickness (Metric) Wall | 1.5mm |