Learn More
Quickfit™ Expansion Socket Adapter
Product Code. 12495436
Change view
Click to view available options
Cone Size:
10/19
14/23
19/26
24/29
29/32
Standard Taper Joint No.:
14/23
19/26
24/29
29/32
34/35
40/38
Unit Size:
Each
Pack of 2
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cone Size Standard Taper Joint No. unitSize
12495436 14/23 24/29 Pack of 2
12146220 14/23 19/26 Pack of 2
10427820 19/26 34/35 Pack of 2
12156220 19/26 24/29 Pack of 2
11928044 14/23 29/32 Pack of 2
12614901 10/19 14/23 Pack of 2
11996124 19/26 29/32 Pack of 2
10881391 24/29 29/32 Pack of 2
12425446 29/32 34/35 Pack of 2
11389093 29/32 40/38 Each
10146454 24/29 34/35 Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 12495436 Supplier Quickfit Supplier No. XA31
Only null left
Add to basket
Add to basket
Made of borosilicate glass. Connects large sized ground glass joints with smaller sized ones. With the QuickFit™ expansion socket adaptor.
Connect large sized ground glass joints with smaller sized ones with the QuickFit™ expansion socket adaptor
  • Useful for the connection of dissimilar sized ground glass joints
  • Large sized socket at the top and small sized cone at the base
TRUSTED_SUSTAINABILITY
Specifications
Cone Size 14/23
For Use With (Application) Useful for the connection of dissimilar sized ground Glass joints
Material Borosilicate Glass
Product Type Expansion adapter
Standard Taper Joint No. 24/29