Learn More
Quickfit™ Distillation Splash Head Adapter
Recieve distillate from a condenser. The QuickFit™ distillation splash head reduces the risk of raw liquid entering into the vapor stream.
85.90 EUR - 99.05 EUR
Specifications
Product Type | Distillation Splash Head Adapter |
---|---|
Material | Borosilicate Glass |
For Use With (Application) | Distillation assemblies in which the condenser is assembled in the vertical mode |
Product Code | Brand | Cone Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cone Size | Price | Quantity & Availability | |||||
12078493
|
Quickfit
SH7/12 |
19/26 (Condenser), 19/26 (flask) |
95.85 EUR
Each |
Estimated Shipment: 04-07-2024
Log in to see stock. |
|||||
12415396
|
Quickfit
SH7/13 |
19/26 (Condenser), 24/29 (flask) |
99.05 EUR
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
12435396
|
Quickfit
SH7/53 |
24/29 (Condenser), 24/29 (flask) |
85.90 EUR
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
Description
Recieve distillate from a condenser. The QuickFit™ distillation splash head reduces the risk of raw liquid entering into the vapor stream.
A splash head for use in distillation assemblies in which the condenser is assembled in the vertical mode - Lower cone accepts flask, upper cone accepts condenser - Splash head reduces entrainment of raw liquid into the vapour streamSpecifications
Distillation Splash Head Adapter | |
Borosilicate Glass | |
Distillation assemblies in which the condenser is assembled in the vertical mode |