Learn More
Quickfit™ Borosilicate Glass Florentine Flask
Product Code. p-8002637
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Pack of 2
Pack of 5
Product Code. 11351895
Brand: Quickfit FD100/3
Only null left
Add to basket
Add to basket
Allows easy removal of solid residues
  • Ideal for use with rotary evaporators
  • Large surface area for rapid evaporation
  • Robust heavy wall construction for added strength
  • Shape allows easy removal of solid residues
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 100 mL
Material Borosilicate Glass
Height (Metric) 108 mm