Learn More
Quickfit™ Borosilicate Glass Cylindrical Culture Vessel
Product Code. p-8002631
Click to view available options
Diameter (Metric):
80 mm
105 mm
133 mm
181 mm
227 mm
288 mm
Capacity (Metric):
10,000 mL
1000 mL
20,000 mL
2000 mL
500 mL
5000 mL
Height (Metric):
130 mm
160 mm
225 mm
290 mm
375 mm
465 mm
Unit Size:
Each
Pack of 5
Product Code. 11371995
Brand: Quickfit FV1L
Manufactured from temperature resistant borosilicate glass. Quickfit™ Borosilicate Glass Cylindrical Culture Vessel is ideal for fermentation and culture work. Autoclavable with precision ground flat-flange joint. For use with Quickfit™ flat-flange lids and retaining clips.
Manufactured from temperature resistant borosilicate glass. QuickfitTM Borosilicate Glass Cylindrical Culture Vessel is ideal for fermentation and culture work. Autoclavable with precision ground flat-flange joint. For use with QuickfitTM flat-flange lids and retaining clips.
  • Ideal for fermentation and culture work
  • Manufactured from temperature resistant borosilicate glass
  • Autoclavable
  • With precision ground flat-flange joint
  • For use with Quickfit™ flat-flange lids and retaining clips
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Bottom Shape Flat
Capacity (Metric) 1000 mL
Material Borosilicate Glass
Surface Treatment Cell Culture
Type Culture Vessel
No. per Pack 1 Each
Height (Metric) 160 mm
Diameter (Metric) 105 mm