Learn More
Thermo Scientific Chemicals Quartz tubing, fused, 12mm (0.472in) OD, 8mm (0.315in) ID
Product Code. 15439189
Click to view available options
Diameter (Metric) Outer:
6.35 mm
12 mm
12.7 mm
19 mm
25 mm
Overall Length (Metric):
30.4 cm
122 cm
Unit Size:
120cm
30cm
Product Code. 15439189
Brand: Thermo Scientific Chemicals 042275.VB
Request Bulk or Custom Format
This item requires a price update prior to being ordered.
Request a Price for this item here.
Manufactured by fusing naturally occurring crystalline silica
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Material Fused Quartz
Diameter (English) Outer 0.25 in.
Diameter (Metric) Outer 6.35 mm
Overall Length (Metric) 122 cm
Product Type Fused Quartz Tubing