Learn More
Applied Biosystems™ QuantStudio 6 PRO 0.2mL, 96-well package
GREENER_CHOICE
Product Code. p-7161535 Shop All Applied Biosystems Products
Click to view available options
Warranty:
1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance)
Unit Size:
Each
Content And Storage:
PC (Laptop)
PC (Tower)
Product Code. 16267631
Brand: Applied Biosystems™ A44288
The Applied Biosystems QuantStudio 6 Pro Real-Time PCR System adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint. Offers facial authentication, voice commands, Smart Help, multiplex capability, motorized interchangeable blocks, simplified workflow, intuitive software, and large capacitive touch-screen interface.
The Applied Biosystems QuantStudio 6 PRO Real-Time PCR System has all of the features of the QS6 FLEX system wth added smart features to deliver a smart, innovative and lab-changing workflow experience, including:
  • Facial authentication; Hands-free login and access to your personal profile
  • Hands-free operation; Perform basic operations using voice commands
  • Radio-frequency identification (RFID) smart plate workflow; 2 embedded RFID readers to detect and decode Applied Biosystems™ TaqMan™ Smart Plates
  • Smart Help; For faster, more efficient, and more effective resolution of instrument and application issues
  • Smart Remote Support; A real-time audio/video collaboration tool and an advanced remote desktop support tool that reduces instrument downtime from days to minute
  • Exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation.
  • Designed for ultimate ease-of-use, the system offers high quality, excellent reliability, and an optimal user experience for researchers who want to work smarter in the lab.
TRUSTED_SUSTAINABILITY
Specifications
Block Format Interchangeable
Format 96-well Plate
For Use With (Equipment) QuantStudio™, ViiA™ 6 System
Product Line QuantStudio™ 6 PRO
Display Type Touchscreen
Green Features Energy Efficient, Fewer Resources used and Less Waste
Warranty 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance)
Shipping Condition Room Temperature