Description
Allows for the quick and easy determination of sulfate in solutions. Macherey Nagel™ Quantofix™ Sulfate Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
- Rapid: provides reliable results within two minutes
- Excellent for use in the beverage industry
- Precise: colour charts adjusted and checked using certified standard solutions
- Shelf life: 2.5 years, store below 30°C
Specifications
Specifications
| Color Indicator | Red to Yellow |
| Format | Strip |
| For Use With (Application) | Determination of Sulfate |
| Graduations | <200, >400, >800, >1200, >1600mg/L SO42- |
| Quantity | 100 Pack |
| Storage Requirements | 30°C |
| Test Time | 2 min. |
| Type | Semi-qualitative Test Strip |