missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Quantikine Immunoassay Control Group 4
Description
- For use as quantitative controls for the determination of cytokine concentrations in biological fluids
- Concentrations have been assigned using R&D Systems Quantikine™ kits
- Controls are prepared in diluted porcine serum with preservatives
- They contain recombinant human cytokines at low, medium and high concentrations
- Controls are supplied lyophilized
Specifications
Specifications
| Product Type | Quantikine Immunoassay Control Group 4 |
| For Use With (Application) | Control |
| Quantity | 1 Each |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?