Learn More
Thermo Scientific™ Quanti-Cult™ Pseudomonas paraeruginosa ATCC™ 9027™
Product Code. 12607699
Click to view available options
Description:
Pseudomonas paraeruginosa ATCC™ 9027™
Unit Size:
Each
Product Code. 12607699
Brand: Thermo Scientific™ R4735210
Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.
Thermo Scientific™ Quanti-Cult™ Pseudomonas paraeruginosa ATCC™ 9027™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial, ready for rehydration and use in quality control procedures.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.3mL inoculum
Ordering Alerts:
Yield is 1 Test/reconstituted vial, 10 Tests/kit
Includes:
10 preserved organism vials and 10 rehydration vials.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 9027TM*
Description Pseudomonas paraeruginosa ATCC™ 9027™
Format Quanti-Cult
Product Type Bactidisk
Target Organism Class Gram-negative Rods
Test Type Quantitative QC
Packaging Type 10 Test/Kit
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.