missing translation for 'onlineSavingsMsg'
Learn More

QNZ, MedChemExpress

Product Code. 16412027
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16412027 50 mg 50mg
16418527 5 mg 5mg
16405878 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16412027 Supplier Medchem Express Supplier No. HY13812

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress QNZ (EVP4593) shows strong inhibitory effects on NF-κB transcriptional activation and TNF-α production with IC50s of 11 and 7 nM, respectively. QNZ (EVP4593) is a neuroprotective inhibitor of SOC channel.

QNZ (EVP4593) shows strong inhibitory effects on NF-κB transcriptional activation and TNF-α production with IC50s of 11 and 7 nM, respectively. QNZ (EVP4593) is a neuroprotective inhibitor of SOC channel.

Specifications

Chemical Name or Material QNZ
CAS 545380-34-5
Color Green
Purity Grade Notes Research
Molecular Formula C22H20N4O
Quantity 50 mg
Synonym EVP4593
Solubility Information DMSO : ≥ 37 mg/mL (103.81 mM)
SMILES NC1=CC2=C(NCCC3=CC=C(OC4=CC=CC=C4)C=C3)N=CN=C2C=C1
Molecular Weight (g/mol) 356.42
Formula Weight 356.42
Percent Purity 99.51%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.