missing translation for 'onlineSavingsMsg'
Learn More

Pyrotinib, MedChemExpress

Product Code. 16463948
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463948 10 mg 10mg
16452926 25 mg 25mg
16451049 1 mg 1mg
16446667 5 mg 5mg
16420307 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16463948 Supplier Medchem Express Supplier No. HY104065

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pyrotinib (SHR-1258) is a potent and selective EGFR/HER2 dual inhibitor with IC50s of 13 and 38 nM, respectively.

Pyrotinib (SHR-1258) is a potent and selective EGFR/HER2 dual inhibitor with IC50s of 13 and 38 nM, respectively.

Specifications

Chemical Name or Material Pyrotinib
CAS 1269662-73-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C32H31ClN6O3
Quantity 10 mg
Synonym SHR-1258
Solubility Information DMSO : 10 mg/mL (17.15 mM; ultrasonic and adjust pH to 6 with HCl)
SMILES O=C(NC1=C(OCC)C=C2N=CC(C#N)=C(NC3=CC=C(OCC4=NC=CC=C4)C(Cl)=C3)C2=C1)/C=C/[C@@H]5N(C)CCC5
Molecular Weight (g/mol) 583.08
Formula Weight 583.08
Percent Purity 98.84%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.