missing translation for 'onlineSavingsMsg'
Learn More

Pyronin Y

Product Code. p-7042020
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 15433729

Brand: Thermo Scientific Alfa Aesar J61068.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Marker dye for RNA useful in acid buffer systems | CAS: 92-32-0 | C17H19ClN2O | 302.802 g/mol

Pyronin Y is a cationic dye that intercalates RNA and is visible as a red band during electrophoresis. It has also been shown to accumulate in mitochondria of viable cells. In conjuction with Methyl Green or Nancy-520, Pyronin Y can be used as a fluorescent differential stain of nucleic acids in paraffin tissue sections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyronin Y is a cationic dye that intercalates RNA and is visible as a red band during electrophoresis. It has also been shown to accumulate in mitochondria of viable cells. In conjuction with Methyl Green or Nancy-520, Pyronin Y can be used as a fluorescent differential stain of nucleic acids in paraffin tissue sections.

Solubility
Soluble in water. Also soluble in methyl cellosolve (9 mg/mL) and ethanol (3 mg/mL).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-32-0
Molecular Formula C17H19ClN2O
Molecular Weight (g/mol) 302.802
MDL Number MFCD00011725
InChI Key INCIMLINXXICKS-UHFFFAOYSA-M
Synonym C.I. 45005; Pyronin G
PubChem CID 7085
ChEBI CHEBI:87347
IUPAC Name [6-(dimethylamino)xanthen-3-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC2=C(C=C1)C=C3C=CC(=[N+](C)C)C=C3O2.[Cl-]

Specifications

Melting Point 250°C to 260°C
Beilstein 3795789
Merck Index 14,8007
Solubility Information Soluble in water. Also soluble in methyl cellosolve (9mg/mL) and ethanol (3mg/mL).
Formula Weight 302.8
Color Green
Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material Pyronin Y
compliance-icons
Product Identifier
  • Pyronin Y
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Pyronin Y

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.