Learn More
Pyrimidine-2-carboxamidoxime, 97%, Thermo Scientific™
Product Code. 15490668
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 15490668
Brand: Thermo Scientific Alfa Aesar H54351.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 90993-49-0
Molecular Formula C5H6N4O
Molecular Weight (g/mol) 138.13
MDL Number MFCD09751609,MFCD12093698
InChI Key RYYZBKPHKBOVSQ-UHFFFAOYSA-N
Synonym n-hydroxy-2-pyrimidinecarboximidamide, n-hydroxypyrimidine-2-carboximidamide, n'-hydroxypyrimidine-2-carboximidamide, pyrimidine-2-carboxamide oxime, z-n'-hydroxypyrimidine-2-carboximidamide, 2-pyrimidinecarboxamide oxime, mtmydzfbfhzrij-uhfffaoysa-n, n-hydroxy pyrimidine-carboxamidine, n-hydroxy-pyrimidine-2-carboxamidine, e-n'-hydroxypyrimidine-2-carboximidamide
PubChem CID 51550622
IUPAC Name nitroso(1H-pyrimidin-2-ylidene)methanamine
SMILES NC(N=O)=C1NC=CC=N1
Specifications
Melting Point 230°C to 235°C
Quantity 250 mg
Formula Weight 138.13
Percent Purity 97%
Chemical Name or Material Pyrimidine-2-carboxamidoxime