missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxamine dihydrochloride, Cell Culture Reagent

Product Code. p-7076593
Click to view available options
Quantity:
2.5 g
5 g
10 g
Unit Size:
10g
2.5g
5g
This item is not returnable. View return policy

Product Code. 15480257

Brand: Thermo Scientific Alfa Aesar J62679.27

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 524-36-7 | C8H14Cl2N2O2 | 241.112 g/mol

A vitamin in the Vitamin B-6 family

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 524-36-7
Molecular Formula C8H14Cl2N2O2
Molecular Weight (g/mol) 241.112
MDL Number MFCD00012808
InChI Key HNWCOANXZNKMLR-UHFFFAOYSA-N
Synonym pyridoxamine dihydrochloride, 4-aminomethyl-5-hydroxymethyl-2-methylpyridin-3-ol dihydrochloride, pyridoxamine dichlorohydrate, pyridoxylamine dihydrochloride, 4-aminomethyl-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride, unii-yq8ncr7v0o, pyridoxamine 2hcl, yq8ncr7v0o, 2-methyl-3-hydroxy-4-aminomethyl-5-hydroxymethylpyridene dihydrochloride, pyridoxemine, dihydrochloride
PubChem CID 10664
IUPAC Name 4-(aminomethyl)-5-(hydroxymethyl)-2-methylpyridin-3-ol;dihydrochloride
SMILES CC1=NC=C(C(=C1O)CN)CO.Cl.Cl

Specifications

Melting Point 220°C to 224°C
Packaging Glass bottle
Quantity 2.5 g
Beilstein 3632748
Solubility Information Soluble in DMSO,water,or methanol /n
Formula Weight 241.12
Physical Form Solid
Chemical Name or Material Pyridoxamine dihydrochloride
compliance-icons
Product Identifier
  • Pyridoxamine dihydrochloride, Cell Culture Reagent
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Pyridoxamine dihydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.