missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Pyridoxal HCl
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- CAS Number: 65-22-5
- Molecular Formula: C8H9NO3 HCl
- Molecular Weight: 203.62 g/mol
Specifications
Specifications
| Chemical Name or Material | Pyridoxal HCl |
| CAS | 65-22-5 |
| Quantity | 100 g |
| Molecular Formula | C8H9NO3-HCl |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction