missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-d5, TRC

Product Code. 30427101
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
1g
Product Code. Quantity unitSize
30427101 1 g 1g
30409033 10 g -
30415964 5 g -
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30427101

Brand: Toronto Research Chemicals TRCP9912371G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7291-22-7
Formula Weight 84.0736
InChI Formula InChI=1S/C5H5N/c1-2-4-6-5-3-1/h1-5H/i1D,2D,3D,4D,5D
IUPAC Name 2,3,4,5,6-pentadeuteriopyridine
Molecular Formula C5 2H5 N
Molecular Weight (g/mol) 84.13
SMILES [2H]c1nc([2H])c([2H])c([2H])c1[2H]
Synonym Pyridine-2,3,4,5,6-d5, Pyridine D5 (6CI,7CI,8CI,9CI), Pentadeuteropyridine, Perdeuteriopyridine
Chemical Name or Material Pyridine D5

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.