Learn More
Pyridine-4-carboxamidoxime N-oxide, 97%, Thermo Scientific™
Product Code. 15498777
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15498777 5 g 5g
15488777 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15498777 Supplier Thermo Scientific Alfa Aesar Supplier No. H52205.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 66168-50-1
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.141
MDL Number MFCD00206512
InChI Key DYPGYCQXSLMOIR-UHFFFAOYSA-N
Synonym n-hydroxy-1-oxy-isonicotinamidine, pyridine-4-carboxamidoxime n-oxide, picolinamidoxime n-oxide, 4-amidoximopyridine n-oxide, 4-n'-hydroxycarbamimidoyl pyridine 1-oxide, 4-amino nitroso methylidene pyridin-1 4h-ol, 4-amino nitroso methylidene pyridin-1-ol, 4-n-hydroxycarbamimidoyl pyridin-1-ium-1-olate, 4-n'-hydroxycarbamimidoyl pyridin-1-ium-1-olate, 4-e-n'-hydroxycarbamimidoyl pyridin-1-ium-1-olate
PubChem CID 44613007
IUPAC Name (1-hydroxypyridin-4-ylidene)-nitrosomethanamine
SMILES C1=CN(C=CC1=C(N)N=O)O
Specifications
Quantity 5 g
Formula Weight 153.14
Percent Purity 97%
Chemical Name or Material Pyridine-4-carboxamidoxime N-oxide