missing translation for 'onlineSavingsMsg'
Learn More

Pyridazine-4-carboxylic acid, 97%

Product Code. 11302269
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302269 5 g 5g
11392259 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302269 Supplier Thermo Scientific Alfa Aesar Supplier No. H27246.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 50681-25-9 | C5H4N2O2 | 124.099 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50681-25-9
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.099
MDL Number MFCD00011576
InChI Key JUSIWJONLKBPDU-UHFFFAOYSA-N
Synonym 4-pyridazinecarboxylic acid, 4-carboxypyridazine, 4-pyridazinecarboxylicacid, pubchem9501, acmc-1alr8, pyridazin-4-carboxylic acid, 4-pyridazine-carboxylic acid, ksc269i2f, pyridazin-4-yl formate, 1,2-diazine-4-carboxylic acid
PubChem CID 2761046
IUPAC Name pyridazine-4-carboxylic acid
SMILES C1=CN=NC=C1C(=O)O

Specifications

Melting Point ∽244°C (decomposition)
Quantity 5 g
Formula Weight 124.1
Percent Purity 97%
Chemical Name or Material Pyridazine-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.