Learn More
Corning™ PYREX™ Narrow Mouth Heavy-Duty Glass Erlenmeyer Flask
GREENER_CHOICE
Product Code. p-124500
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
125 mL
250 mL
500 mL
1 L
2000 mL
4 L
6 L
Includes:
No Stopper
Stopper
Unit Size:
Each
Pack of 12
Pack of 6
Product Code. 12580056
Brand: Corning™ undefined
Only null left
Add to basket
Add to basket
Constructed of PYREX™ for durability. Erlenmeyer style, rugged tooled top finish and narrow mouth.
Use these flasks for a variety of applications. PYREX™ Narrow Neck Heavy-Duty Erlenmeyer Flasks are designed for longer life and increased user safety.
TRUSTED_SUSTAINABILITY
Specifications
Description Top style: Reinforced tooled
Diameter (English) Outer 1.61 in.
Capacity (Metric) 25 mL
Material Borosilicate Glass
Graduation Range 10 to 25 mL
Graduations 5 mL
Height (English) 2.55 in.
Height (Metric) 6.5 cm
Includes No Stopper
Neck Style Narrow Mouth
No. per Case 48/Cs.
No. per Pack 12/Pk.
Writing Surface Large Enamel Marking
Disposable Reusable
Mouth Style Narrow
Sterility Non-sterile
Diameter (Metric) Outer 41 mm
Closure Size 0