Learn More
Pyrex™ Borosilicate Glass Round Bottom Culture Tube with Screw Cap
Product Code. 11377814
Click to view available options
Capacity (Metric):
9 mL
11 mL
14 mL
15 mL
19 mL
20 mL
28 mL
36 mL
40 mL
50 mL
70 mL
Length (Metric):
100 mm
125 mm
160 mm
180 mm
200 mm
Unit Size:
Each
Pack of 40
Product Code. 11377814
Brand: Pyrex 1636/24MP
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ideal for culture work with PBT screw caps and PTFE lined rubber disc. Pryex™ culture tubes come in a wide range of sizes to meet your needs.
Ideal for culture work with PBT screw caps and PTFE lined rubber disc. Pryex™ culture tubes come in a wide range of sizes to meet your needs.
  • Ideal for culture work
  • Manufactured from Pyrex™ borosilicate glass
  • High resistance to attack from water reduces leaching of contaminants which can cause pH changes
  • Phenolic screw caps with inert PTFE lined rubber disc
  • Cap can withstand repeated dry heat and steam sterilisation cycles (121°C for two hours)
TRUSTED_SUSTAINABILITY
Specifications
Closure Material Phenolic
Capacity (Metric) 9 mL
Length (Metric) 100 mm
Material Borosilicate Glass
For Use With Ideal for culture work
Includes Screw cap
Liner Material PTFE
Thread Size 13 mm
Autoclavable Autoclavable
Diameter (Metric) 14 mm
Closure Type Screw Cap